7-Bromo-4-fluoroindole 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-4-fluoro-1H-indole

PC7088-1G PC7088-250MG
APOSPC7088-1GEA 248 EUR
APOSPC7088-1G APOSPC7088-250MG
7-Bromo-4-fluoroindole 98%
7-Bromo-4-fluoroindole
Formel: C₈H₅BrFN
Molecular Weight: 214.04 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04037874
CAS-Nummer: 292636-09-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR