4-Carboxy-3-fluorphenylboronsäurepinakolester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

PC7092-1G PC7092-5G
APOSPC7092-1GEA 66.7 EUR
APOSPC7092-1G APOSPC7092-5G
4-Carboxy-3-fluorphenylboronsäurepinakolester 98%
4-Carboxy-3-fluorphenylboronsäurepinakolester
Formel: C₁₃H₁₆BFO₄
Molecular Weight: 266,0732 g/mol
Schmelzpunkt: 289…295 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027284
CAS-Nummer: 867256-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR