4-(2,2,2-Trifluorethoxy)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(2,2,2-Trifluoroethoxy)phenyl)boronic acid, 4-(2,2,2-Trifluoroethoxy)benzeneboronic acid

PC7094-1G PC7094-5G
APOSPC7094-1GEA 72.5 EUR
APOSPC7094-1G APOSPC7094-5G
4-(2,2,2-Trifluorethoxy)phenylboronsäure 98%
4-(2,2,2-Trifluorethoxy)phenylboronsäure
Formel: C₈H₈BF₃O₃
Molecular Weight: 219,95 g/mol
Schmelzpunkt: 191…194 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09027239
CAS-Nummer: 886536-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR