4-Brom-3-fluorthioanisol 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Brom-2-fluor-4-(methylthio)benzol
PC7095-1G
PC7095-25G
PC7095-5G
APOSPC7095-1GEA
102
EUR
APOSPC7095-1G
APOSPC7095-25G
APOSPC7095-5G
4-Brom-3-fluorthioanisol 96%
4-Brom-3-fluorthioanisol
Formel:
C₇H₆BrFS Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027283 CAS-Nummer: 917562-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...