1-[6-(Trifluoromethyl)pyridin-2-yl]piperazine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC7099-1G
PC7099-5G
PC7099-25G
APOSPC7099-1GEA
71.2
EUR
APOSPC7099-1G
APOSPC7099-5G
APOSPC7099-25G
1-[6-(Trifluoromethyl)pyridin-2-yl]piperazine 97%
1-[6-(Trifluoromethyl)pyridin-2-yl]piperazine
Formel:
C₁₀H₁₂F₃N₃ Siedepunkt: 102 °C/0,5mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00114706 CAS-Nummer: 127561-18-6 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...