2-Fluorisonicotinonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluor-4-pyridincarbonsäurenitril, 4-Cyan-2-fluorpyridin
PC7107-5G
PC7107-25G
PC7107-100G
APOSPC7107-5GEA
36.4
EUR
APOSPC7107-5G
APOSPC7107-25G
APOSPC7107-100G
2-Fluorisonicotinonitril 98%
2-Fluorisonicotinonitril
Formel:
C₆H₃FN₂ Molecular Weight: 122,1 g/mol Schmelzpunkt: 29…34 °C Flammpunkt: 82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027299 CAS-Nummer: 3939-14-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...