5-Chlor-2-fluorphenylhydrazin Hydrochlorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Chloro-2-fluorophenyl)hydrazine hydrochloride

PC7116-1G PC7116-5G PC7116-25G
APOSPC7116-1GEA 21.9 EUR
APOSPC7116-1G APOSPC7116-5G APOSPC7116-25G
5-Chlor-2-fluorphenylhydrazin Hydrochlorid 97%
5-Chlor-2-fluorphenylhydrazin Hydrochlorid
Formel: C₆H₇Cl₂FN₂
Molecular Weight: 197,0376 g/mol
Schmelzpunkt: 205…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094414
CAS-Nummer: 529512-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR