2-Fluor-4-biphenylboronsäurepinakolester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2-Fluoro-[1,1'-biphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PC7129-100MG
PC7129-1G
PC7129-5G
APOSPC7129-100MGEA
21.9
EUR
APOSPC7129-100MG
APOSPC7129-1G
APOSPC7129-5G
2-Fluor-4-biphenylboronsäurepinakolester 98%
2-Fluor-4-biphenylboronsäurepinakolester
Formel:
C₁₈H₂₀BFO₂ Molecular Weight: 298,160 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09258651 CAS-Nummer: 269410-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...