3-Brom-6-fluor-2-methylpyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Brom-2-fluor-6-picolin

PC7144-1G PC7144-5G PC7144-25G
APOSPC7144-1GEA 21.9 EUR
APOSPC7144-1G APOSPC7144-5G APOSPC7144-25G
3-Brom-6-fluor-2-methylpyridin 98%
3-Brom-6-fluor-2-methylpyridin
Formel: C₆H₅BrFN
Molecular Weight: 190.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095092
CAS-Nummer: 375368-83-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR