2-Fluor-3-(trifluormethyl)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-3-(trifluoromethyl)benzeneboronic acid

PC7181-1G PC7181-5G PC7181-10G
APOSPC7181-1GEA 21.9 EUR
APOSPC7181-1G APOSPC7181-5G APOSPC7181-10G
2-Fluor-3-(trifluormethyl)phenylboronsäure 97%
2-Fluor-3-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BF₄O₂
Molecular Weight: 207,92 g/mol
Schmelzpunkt: 60…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07784336
CAS-Nummer: 157834-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR