2',3',6'-Trifluoracetophenon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2,3,6-Trifluorphenyl)ethan-1-on

PC7202G-1G PC7202G-5G PC7202G-10G
APOSPC7202G-1GEA 21.9 EUR
APOSPC7202G-1G APOSPC7202G-5G APOSPC7202G-10G
2',3',6'-Trifluoracetophenon 97%
2',3',6'-Trifluoracetophenon
Formel: C₈H₅F₃O
Molecular Weight: 174,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070809
CAS-Nummer: 208173-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR