5-Brom-2-chlor-3-fluorpyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC7206-5G
APOSPC7206-5GEA
116
EUR
APOSPC7206-5G
5-Brom-2-chlor-3-fluorpyridin 98%
5-Brom-2-chlor-3-fluorpyridin
Formel:
C₅H₂BrClFN Molecular Weight: 210,43 g/mol Schmelzpunkt: 50…52 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03411574 CAS-Nummer: 831203-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...