3-Amino-5-brom-2-fluorpyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-fluoropyridin-3-amine

PC7207-5G PC7207-10G
APOSPC7207-5GEA 143 EUR
APOSPC7207-5G APOSPC7207-10G
3-Amino-5-brom-2-fluorpyridin
3-Amino-5-bromo-2-fluoropyridine
Formel: C₅H₄BrFN₂
Molecular Weight: 191.0011 g/mol
Schmelzpunkt: 75…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659524
CAS-Nummer: 884495-22-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR