N-[3-(Trifluoromethyl)pyrid-2-yl]-N-methyl-hydrazine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(N-Methylhydrazino)-3-(trifluoromethyl)pyridine
PC7221-1G
PC7221-250MG
PC7221-500MG
APOSPC7221-1GEA
0
EUR
APOSPC7221-1G
APOSPC7221-250MG
APOSPC7221-500MG
N-[3-(Trifluoromethyl)pyrid-2-yl]-N-methyl-hydrazine
N-[3-(Trifluoromethyl)pyrid-2-yl]-N-methyl-hydrazine
Formel:
C₇H₈F₃N₃ Molecular Weight: 191,16 g/mol Siedepunkt: 90 °C/15mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD00052298 CAS-Nummer: 175205-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...