1-(Trifluoracetyl)imidazol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,2-Trifluoro-1-(1H-imidazol-1-yl)ethanone

PC7240-25G PC7240-5G
APOSPC7240-25GEA 102 EUR
APOSPC7240-25G APOSPC7240-5G
1-(Trifluoracetyl)imidazol 95%
1-(Trifluoracetyl)imidazol

Derivatising agent. Useful for trifluoroacetylation ofalcohols and amines for electron-capture GC analysis.

Formel: C₅H₃F₃N₂O
Molecular Weight: 164.09 g/mol
Siedepunkt: 137…138 °C
Dichte: 1.441 g/cm³ (25 °C)
Flammpunkt: 45 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00014501
CAS-Nummer: 1546-79-8
EINECS: 216-282-5
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR