Methyl-3-fluor-4-nitrobenzoat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluor-4-nitrobenzoesäuremethylester

PC7249-1G PC7249-25G PC7249-5G
APOSPC7249-1GEA 21.9 EUR
APOSPC7249-1G APOSPC7249-25G APOSPC7249-5G
Methyl-3-fluor-4-nitrobenzoat 98%
Methyl-3-fluor-4-nitrobenzoat
Formel: C₈H₆FNO₄
Molecular Weight: 199,14 g/mol
Schmelzpunkt: 53…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08444027
CAS-Nummer: 185629-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR