2-Brom-6-fluornaphthalin
Lieferant: Apollo Scientific
|
Achtung
|
PC7265-1G
PC7265-5G
APOSPC7265-1GEA
55.1
EUR
APOSPC7265-1G
APOSPC7265-5G
2-Brom-6-fluornaphthalin
2-Bromo-6-fluoronaphthalene
Formel:
C₁₀H₆BrF Molecular Weight: 225,0600 g/mol Schmelzpunkt: 66…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09763683 CAS-Nummer: 324-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...