2,3,4-Trifluorbenzamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3,4-Trifluorbenzoesäureamid

PC7265T-1G PC7265T-25G PC7265T-5G
APOSPC7265T-1GEA 21.9 EUR
APOSPC7265T-1G APOSPC7265T-25G APOSPC7265T-5G
2,3,4-Trifluorbenzamid 97%
2,3,4-Trifluorbenzamid
Formel: C₇H₄F₃NO
Molecular Weight: 175.11 g/mol
Schmelzpunkt: 127…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061265
CAS-Nummer: 207919-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR