2,4,6-Trifluorbenzamid 97%

Lieferant: Apollo Scientific
Achtung

PC7265X-10G PC7265X-5G
APOSPC7265X-10GEA 109 EUR
APOSPC7265X-10G APOSPC7265X-5G
2,4,6-Trifluorbenzamid 97%
2,4,6-Trifluorbenzamid
Formel: C₇H₄F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00061199
CAS-Nummer: 82019-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR