3,4,5-Trifluorbenzamid

Lieferant: Apollo Scientific
Achtung

PC7265Y-25G PC7265Y-5G PC7265Y-1G
APOSPC7265Y-25GEA 343 EUR
APOSPC7265Y-25G APOSPC7265Y-5G APOSPC7265Y-1G
3,4,5-Trifluorbenzamid
3,4,5-Trifluorbenzamid
Formel: C₇H₄F₃NO
Molecular Weight: 175,11 g/mol
Schmelzpunkt: 144…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00083524
CAS-Nummer: 78324-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR