3-Fluor-4-(propylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Fluoro-4-(propylcarbamoyl)phenyl)boronic acid

PC7267-1G PC7267-5G
APOSPC7267-1GEA 224 EUR
APOSPC7267-1G APOSPC7267-5G
3-Fluor-4-(propylcarbamoyl)phenylboronsäure 98%
3-Fluor-4-(propylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₃BFNO₃
Molecular Weight: 225.0300 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09258737
CAS-Nummer: 874289-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR