2,3,4-Trifluorbenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
PC7281L-100G
PC7281L-250G
PC7281L-500G
APOSPC7281L-100GEA
56.7
EUR
APOSPC7281L-100G
APOSPC7281L-250G
APOSPC7281L-500G
2,3,4-Trifluorbenzoesäure
2,3,4-Trifluorobenzoic acid
Formel:
C₇H₃F₃O₂ Molecular Weight: 176.09 g/mol Siedepunkt: 237 Schmelzpunkt: 140…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061232 CAS-Nummer: 61079-72-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...