3,4,5-Trifluorbenzophenon 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(Phenylmethanoyl)-3,4,5-trifluorbenzene
PC7284I-1G
PC7284I-5G
PC7284I-25G
APOSPC7284I-1GEA
29
EUR
APOSPC7284I-1G
APOSPC7284I-5G
APOSPC7284I-25G
3,4,5-Trifluorbenzophenon 97%
3,4,5-Trifluorbenzophenon
Formel:
C₁₃H₇F₃O Molecular Weight: 236.19 g/mol Schmelzpunkt: 77…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00083525 CAS-Nummer: 70028-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...