2,3,4-Trifluorbenzylalkohol 97%

Lieferant: Apollo Scientific
Achtung

PC7285L-25G PC7285L-5G
APOSPC7285L-25GEA 115 EUR
APOSPC7285L-25G APOSPC7285L-5G
2,3,4-Trifluorbenzylalkohol 97%
2,3,4-Trifluorbenzylalkohol
Formel: C₇H₅F₃O
Molecular Weight: 162,11 g/mol
Schmelzpunkt: 29,6 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00083551
CAS-Nummer: 144284-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR