2,4,6-Trifluorbenzylalkohol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC7285LF-10G
PC7285LF-25G
APOSPC7285LF-10GEA
55.1
EUR
APOSPC7285LF-10G
APOSPC7285LF-25G
2,4,6-Trifluorbenzylalkohol 97%
2,4,6-Trifluorbenzylalkohol
Formel:
C₇H₅F₃O Molecular Weight: 162.11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061207 CAS-Nummer: 118289-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...