1-Brom-4,4,4-trifluorbutan 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC7288-25G
PC7288-5G
PC7288-1G
APOSPC7288-25GEA
364
EUR
APOSPC7288-25G
APOSPC7288-5G
APOSPC7288-1G
1-Brom-4,4,4-trifluorbutan 98%
1-Bromo-4,4,4-trifluorobutane
Formel:
C₄H₆BrF₃ Molecular Weight: 190,99 g/mol Siedepunkt: 103…105 °C Dichte: 1,555 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066335 CAS-Nummer: 406-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...