1,1,1-Trifluoro-2-butene 97%

Lieferant: Apollo Scientific
Achtung

PC7291F-5G PC7291F-25G
APOSPC7291F-5GEA 273 EUR
APOSPC7291F-5G APOSPC7291F-25G
1,1,1-Trifluoro-2-butene 97%
1,1,1-Trifluoro-2-butene
Formel: C₄H₅F₃
Molecular Weight: 110,08 g/mol
Siedepunkt: 17 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082625
CAS-Nummer: 406-39-3
UN: 3161
ADR: 2.1,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR