2-Fluor-6-methoxybenzamid

Lieferant: Apollo Scientific
Achtung

PC7323-1G PC7323-5G
APOSPC7323-1GEA 154 EUR
APOSPC7323-1G APOSPC7323-5G
2-Fluor-6-methoxybenzamid
2-Fluor-6-methoxybenzamid
Formel: C₈H₈FNO₂
Molecular Weight: 169.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00277474
CAS-Nummer: 529512-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR