4-Chlor-8-fluorchinolin

Lieferant: Apollo Scientific
Achtung

PC7341-1G PC7341-5G
APOSPC7341-1GEA 40.6 EUR
APOSPC7341-1G APOSPC7341-5G
4-Chlor-8-fluorchinolin
4-Chloro-8-fluoroquinoline
Formel: C₉H₅ClFN
Molecular Weight: 181,60 g/mol
Schmelzpunkt: 90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00278785
CAS-Nummer: 63010-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR