4-Diethylamino-1,1,1-trifluorobut-3-en-2-one

Lieferant: Apollo Scientific
Achtung

PC7356-10G PC7356-1G PC7356-5G
APOSPC7356-10GEA 517 EUR
APOSPC7356-10G APOSPC7356-1G APOSPC7356-5G
4-Diethylamino-1,1,1-trifluorobut-3-en-2-one
4-Diethylamino-1,1,1-trifluorobut-3-en-2-one
Formel: C₈H₁₂F₃NO
Molecular Weight: 195,19 g/mol
Siedepunkt: 97…98 °C/ 2mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00115108
CAS-Nummer: 21045-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR