4'-Brom-3'-fluoracetanilid

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(4-Brom-3-fluorphenyl)acetamid

PC7364-25G PC7364-5G PC7364-1G
APOSPC7364-25GEA 238 EUR
APOSPC7364-25G APOSPC7364-5G APOSPC7364-1G
4'-Brom-3'-fluoracetanilid
4'-Brom-3'-fluoracetanilid
Formel: H₃CC(O)NH(C₆H₃BrF)
Molecular Weight: 232,05 g/mol
Schmelzpunkt: 149…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00672937
CAS-Nummer: 351-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR