1,1,1-Trifluoriodethan 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2,2-Trifluoriodethan, 2-Iod-1,1,1-trifluorethan, 1-Iod-2,2,2-trifluorethan, 2,2,2-Trifluorethyliodid
PC7370-100G
PC7370-250G
PC7370-25G
APOSPC7370-100GEA
116
EUR
APOSPC7370-100G
APOSPC7370-250G
APOSPC7370-25G
1,1,1-Trifluoriodethan 99%
1,1,1-Trifluoriodethan
Formel:
C₂H₂F₃I Molecular Weight: 209,94 g/mol Siedepunkt: 53…55 °C Dichte: 2,13 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00001082 CAS-Nummer: 353-83-3 EINECS: 206-541-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...