2-Brom-4-fluorbenzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Brom-4-fluorphenyl)methanol

PC7372-25G PC7372-5G
APOSPC7372-25GEA 69.6 EUR
APOSPC7372-25G APOSPC7372-5G
2-Brom-4-fluorbenzylalkohol 98%
2-Brom-4-fluorbenzylalkohol
Formel: C₇H₆BrFO
Molecular Weight: 205,03 g/mol
Schmelzpunkt: 68…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00672925
CAS-Nummer: 229027-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR