2-Brom-5-fluorbenzylalkohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Bromo-5-fluorophenyl)methanol
PC7373-25G
PC7373-5G
APOSPC7373-25GEA
37.7
EUR
APOSPC7373-25G
APOSPC7373-5G
2-Brom-5-fluorbenzylalkohol
2-Brom-5-fluorbenzylalkohol
Formel:
C₇H₆BrFO Molecular Weight: 205,03 g/mol Schmelzpunkt: 91…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142876 CAS-Nummer: 202865-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...