3-Brom-4-fluorbenzylalkohol 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Bromo-4-fluorophenyl)methanol
PC7379-25G
PC7379-5G
APOSPC7379-25GEA
37.7
EUR
APOSPC7379-25G
APOSPC7379-5G
3-Brom-4-fluorbenzylalkohol 99%
3-Brom-4-fluorbenzylalkohol
Formel:
C₇H₆BrFO Molecular Weight: 205,03 g/mol Siedepunkt: 80 °C/0,5mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143093 CAS-Nummer: 77771-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...