4-Brom-2-fluorbenzylalkohol 98+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Brom-2-fluorphenyl)methanol
PC7381-100G
PC7381-25G
PC7381-5G
APOSPC7381-100GEA
108
EUR
APOSPC7381-100G
APOSPC7381-25G
APOSPC7381-5G
4-Brom-2-fluorbenzylalkohol 98+%
4-Brom-2-fluorbenzylalkohol
Formel:
C₇H₆BrFO Molecular Weight: 205,03 g/mol Schmelzpunkt: 30…35 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143265 CAS-Nummer: 188582-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...