2-Bromo-4-fluorobenzylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Bromo-4-fluorophenyl)methanamine hydrochloride

PC7383-5G PC7383-1G PC7383-25G
APOSPC7383-5GEA 132 EUR
APOSPC7383-5G APOSPC7383-1G APOSPC7383-25G
2-Bromo-4-fluorobenzylamine hydrochloride
2-Bromo-4-fluorobenzylamine hydrochloride
Formel: C₇H₈BrClFN
Molecular Weight: 240,5 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672922
CAS-Nummer: 289038-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR