2-Brom-4-fluorbenzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
PC7395-25G
PC7395-5G
APOSPC7395-25GEA
21.9
EUR
APOSPC7395-25G
APOSPC7395-5G
2-Brom-4-fluorbenzaldehyd
2-Bromo-4-fluorobenzaldehyde
Formel:
C₇H₄BrFO Molecular Weight: 203.01 g/mol Schmelzpunkt: 64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672923 CAS-Nummer: 59142-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...