5-Brom-2-iodbenzotrifluorid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Brom-2-(trifluormethyl)iodbenzol, 5-Brom-2-iodbenzotrifluorid, 1-Brom-4-iod-3-(trifluormethyl)benzol

PC7413-100G PC7413-25G PC7413-5G
APOSPC7413-100GEA 121 EUR
APOSPC7413-100G APOSPC7413-25G APOSPC7413-5G
5-Brom-2-iodbenzotrifluorid 98%
5-Brom-2-iodbenzotrifluorid
Formel: C₇H₃BrF₃I
Molecular Weight: 350.9
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00672935
CAS-Nummer: 364-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR