4-Chlor-2-fluoranisol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-2-fluoro-1-methoxybenzene

PC7423-1G PC7423-5G
APOSPC7423-1GEA 21.9 EUR
APOSPC7423-1G APOSPC7423-5G
4-Chlor-2-fluoranisol 98%
4-Chlor-2-fluoranisol
Formel: C₇H₆ClFO
Molecular Weight: 160.58
Lagertemperatur: Raumtemperatur
MDL: MFCD00672964
CAS-Nummer: 452-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR