2-Methyl-4-(trifluormethyl)phenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC7424-1G
PC7424-5G
PC7424-250MG
APOSPC7424-1GEA
154
EUR
APOSPC7424-1G
APOSPC7424-5G
APOSPC7424-250MG
2-Methyl-4-(trifluormethyl)phenylboronsäure 97%
2-Methyl-4-(trifluormethyl)phenylboronsäure
Formel:
C₈H₈BF₃O₂ Molecular Weight: 203,9541 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08689550 CAS-Nummer: 957034-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...