4-Brom-3-methylbenzotrifluorid 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-methyl-4-(trifluoromethyl)benzene
PC7427-1G
PC7427-25G
PC7427-5G
APOSPC7427-1GEA
49.5
EUR
APOSPC7427-1G
APOSPC7427-25G
APOSPC7427-5G
4-Brom-3-methylbenzotrifluorid 96%
4-Brom-3-methylbenzotrifluorid
Formel:
C₈H₆BrF₃ Molecular Weight: 239,0325 g/mol Dichte: 1,54 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258773 CAS-Nummer: 929000-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...