3-Benzyloxy-2,6-difluorphenylboronsäure 98%

Lieferant: Apollo Scientific

PC7428-1G PC7428-5G
APOSPC7428-1GEA 71.2 EUR
APOSPC7428-1G APOSPC7428-5G
3-Benzyloxy-2,6-difluorphenylboronsäure 98%
3-Benzyloxy-2,6-difluorphenylboronsäure
Formel: C₁₃H₁₁BF₂O₃
Molecular Weight: 264.0325 g/mol
Schmelzpunkt: 112…118 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06200705
CAS-Nummer: 870718-07-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR