3-(Trifluormethoxy)benzylalkohol 97%

Lieferant: Apollo Scientific
Achtung

PC7438SC-25G PC7438SC-5G
APOSPC7438SC-25GEA 69.6 EUR
APOSPC7438SC-25G APOSPC7438SC-5G
3-(Trifluormethoxy)benzylalkohol 97%
3-(Trifluormethoxy)benzylalkohol
Formel: C₈H₇F₃O₂
Molecular Weight: 192,14 g/mol
Siedepunkt: 97…98 °C/11mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00061270
CAS-Nummer: 50823-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR