4-(Trifluoromethoxy)phenoxyacetic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-(Trifluoromethoxy)phenoxy)acetic acid
PC7439P-10G
PC7439P-1G
APOSPC7439P-10GEA
101
EUR
APOSPC7439P-10G
APOSPC7439P-1G
4-(Trifluoromethoxy)phenoxyacetic acid 97%
4-(Trifluoromethoxy)phenoxyacetic acid
Formel:
C₉H₇F₃O₄ Molecular Weight: 236,15 g/mol Schmelzpunkt: 84…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052352 CAS-Nummer: 72220-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...