(3-(Trifluormethoxy)phenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Trifluoromethoxy)phenylacetic acid
PC7440E-100G
PC7440E-25G
APOSPC7440E-100GEA
270
EUR
APOSPC7440E-100G
APOSPC7440E-25G
(3-(Trifluormethoxy)phenyl)essigsäure 98%
(3-(Trifluormethoxy)phenyl)essigsäure
Formel:
(C₆H₄(OCF₃))CH₂COOH Molecular Weight: 220,15 g/mol Schmelzpunkt: 53…55 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082480 CAS-Nummer: 203302-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...