(4-(Trifluormethoxy)phenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Trifluoromethoxy)phenylacetic acid
PC7440F-1G
PC7440F-25G
PC7440F-5G
APOSPC7440F-1GEA
21.9
EUR
APOSPC7440F-1G
APOSPC7440F-25G
APOSPC7440F-5G
(4-(Trifluormethoxy)phenyl)essigsäure 98%
(4-(Trifluormethoxy)phenyl)essigsäure
Formel:
(C₆H₄(OCF₃))CH₂COOH Molecular Weight: 220,15 g/mol Schmelzpunkt: 85…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061239 CAS-Nummer: 4315-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...