3-(Trifluormethoxy)phenylacetonitril 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(Trifluormethoxy)benzylcyanid

PC7440G-1G
APOSPC7440G-1GEA 21.9 EUR
APOSPC7440G-1G
3-(Trifluormethoxy)phenylacetonitril 98%
3-(Trifluormethoxy)phenylacetonitril
Formel: C₉H₆F₃NO
Molecular Weight: 201,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061272
CAS-Nummer: 108307-56-8
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR