1-[2-Nitro-4-(trifluoromethyl)phenyl]-4-methylpiperidine

Lieferant: Apollo Scientific

Synonyme: 4-Methyl-1-[2-nitro-4-(trifluoromethyl)phenyl]piperidine

PC7454-10G PC7454-1G PC7454-5G
APOSPC7454-10GEA 303 EUR
APOSPC7454-10G APOSPC7454-1G APOSPC7454-5G
1-[2-Nitro-4-(trifluoromethyl)phenyl]-4-methylpiperidine
1-[2-Nitro-4-(trifluoromethyl)phenyl]-4-methylpiperidine
Formel: C₁₃H₁₅F₃N₂O₂
Schmelzpunkt: 38 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027734
CAS-Nummer: 392237-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR