1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxamide

Lieferant: Apollo Scientific

PC7455-1G PC7455-5G
APOSPC7455-1GEA 111 EUR
APOSPC7455-1G APOSPC7455-5G
1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxamide
1-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxamide
Formel: C₁₃H₁₄F₃N₃O₃
Schmelzpunkt: 177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01418201
CAS-Nummer: 329064-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR